Order Entry
Export
ContactUsLinkComponent
3,5-Bis(trifluoromethyl)chlorobenzene 97%
3,5-Bis(trifluoromethyl)chlorobenzene 97%
Catalog # APOSPC1351-1G
Supplier:  Apollo Scientific
CAS Number:  
3,5-Bis(trifluoromethyl)chlorobenzene 97%
Catalog # APOSPC1351-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :248.553 g/mol
Storage Temperature:Ambient
CAS Number:328-72-3
Formula:C₈H₃ClF₆
MDL Number:MFCD01861848