Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Butyl 2,2-difluoroacetate
Butyl 2,2-difluoroacetate 98%
Butyl 2,2-difluoroacetate 98%
:
Apollo Scientific
Butyl 2,2-difluoroacetate 98%
:
Apollo Scientific
:
368-35-4
:
:
C₆H₁₀F₂O₂
:
MFCD03094108
:
152.139 g/mol
:
137 °C
:
1.327 g/cm³ (25 °C)
:
37 °C
:
Ambient
:
1993
:
3,III
:
GHS02
:
GHS07