

Methyl 3,3-difluoroacrylate
Catalog # APOSPC1244-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:122.07 g/mol
Boiling Pt:82…83 °C
Density:1.199 g/cm³ (25 °C)
Storage Temperature:Refrigerator
UN:1993
ADR:3,III
:GHS02
:GHS07
CAS Number:406-05-3
Formula:C₄H₄F₂O₂
MDL Number:MFCD07772050