Order Entry
Export
ContactUsLinkComponent
 
Flammable
Compressed Gas
1-Bromo-1,2-difluoroethylene
Catalog # APOSPC1243-1G
Supplier:  Apollo Scientific
CAS Number:  
Flammable
Compressed Gas
1-Bromo-1,2-difluoroethylene
Catalog # APOSPC1243-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Cylinder
  • Pk:
    1 g
  • Seq:
    3047
  • Pk Info:
    1/4" NPT connection

Specifications

About this item

 :
 :142.93 g/mol
Boiling Pt:15…20 °C (760 mm Hg)
Density:1.5 g/cm³ (25 °C)
 :GHS02
 :GHS04
Storage Temperature:Ambient
CAS Number:358-99-6
Formula:C₂HBrF₂
MDL Number:MFCD09998062