Order Entry
Export
ContactUsLinkComponent
2-Amino-4,6-difluorobenzotrifluoride 97%
Irritant
2-Amino-4,6-difluorobenzotrifluoride 97%
Catalog # APOSPC10593-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Amino-4,6-difluorobenzotrifluoride 97%
Catalog # APOSPC10593-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :197.105 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:123973-33-1
Formula:C₇H₄F₅N
MDL Number:MFCD04972831