

1,4-Dimethyl-2-fluorobenzene
Catalog # APOSPC10440-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Boiling Pt:137…138 °C
Melting Pt:–6 °C
Flash Pt:39 °C
Storage Temperature:Ambient
ADR:3,III
:124.155 g/mol
UN:1993
:GHS02
:GHS07
CAS Number:696-01-5
Formula:C₈H₉F
MDL Number:MFCD03412243