2-(3,4-Difluorophenoxy)benzenecarboxaldehyde oxime
Catalog # APOSPC10235-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:249.213 g/mol
Melting Pt:73…75 °C
Storage Temperature:Ambient
CAS Number:451485-69-1
Formula:C₁₃H₉F₂NO₂
MDL Number:MFCD02091017