Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Amino-2,6-difluorobenzotrifluoride
4-Amino-2,6-difluorobenzotrifluoride
4-Amino-2,6-difluorobenzotrifluoride
APOSPC0289-5G
:
Apollo Scientific
:
4-Amino-2,6-difluorobenzotrifluoride
APOSPC0289-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
197.105 g/mol
:
103…105 °C
:
66 °C
:
Refrigerator
:
2811
:
6.1,III
:
GHS06
:
123950-44-7
:
C₇H₄F₅N
:
MFCD00190122