Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Dichloro-5-fluorobenzaldehyde
2,4-Dichloro-5-fluorobenzaldehyde
2,4-Dichloro-5-fluorobenzaldehyde
Corrosive
Irritant
2,4-Dichloro-5-fluorobenzaldehyde
Supplier:
Apollo Scientific
Certificates
Expand image
Corrosive
Irritant
2,4-Dichloro-5-fluorobenzaldehyde
Supplier:
Apollo Scientific
About this item
CAS Number:
86522-91-0
:
Formula:
C₇H₃Cl₂FO
MDL Number:
MFCD01631379
:
193.003 g/mol
:
GHS05
:
GHS07
Storage Temperature:
Ambient