1,3-Dimethoxy-1,1,2,3,3-pentafluoropropane
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:758-62-3
:
Formula:C₅H₇F₅O₂
MDL Number:MFCD16170455
:194.1 g/mol
Boiling Pt:64…65 °C
Density:1.3673 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:1993
ADR:3,III