Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,5-Dibromo-2-chloropyridine
4,5-Dibromo-2-chloropyridine ≥97%
4,5-Dibromo-2-chloropyridine ≥97%
4,5-Dibromo-2-chloropyridine ≥97%
APOSOR966917-250MG
:
Apollo Scientific
:
4,5-Dibromo-2-chloropyridine ≥97%
APOSOR966917-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
271.337 g/mol
:
Refrigerator
:
1807166-31-9
:
C₅H₂Br₂ClN
:
MFCD28750474