Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,7-Diazaspiro[4.4]nonane dihydrobromide
2,7-Diazaspiro[4.4]nonane dihydrobromide 95+%
2,7-Diazaspiro[4.4]nonane dihydrobromide 95+%
Irritant
2,7-Diazaspiro[4.4]nonane dihydrobromide 95+%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,7-Diazaspiro[4.4]nonane dihydrobromide 95+%
Supplier:
Apollo Scientific
About this item
CAS Number:
77415-55-5
:
Formula:
C₇H₁₄N₂·2BrH
MDL Number:
MFCD27964259
:
288.023 g/mol
:
GHS07
Storage Temperature:
Ambient