3,4-Dichloroisothiazole-5-carbonitrile 95%
Catalog # APOSOR964430-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:179.027 g/mol
Storage Temperature:Ambient
CAS Number:18480-52-9
Formula:C₄Cl₂N₂S
MDL Number:MFCD09743745