Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(But-3-yn-2-yl)isoindoline-1,3-dione
2-(But-3-yn-2-yl)isoindoline-1,3-dione 95%
2-(But-3-yn-2-yl)isoindoline-1,3-dione 95%
2-(But-3-yn-2-yl)isoindoline-1,3-dione 95%
Supplier:
Apollo Scientific
Certificates
Expand image
2-(But-3-yn-2-yl)isoindoline-1,3-dione 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
14396-89-5
:
Formula:
C₁₂H₉NO₂
MDL Number:
MFCD08703146
:
199.205 g/mol
Storage Temperature:
Ambient