Order Entry
Export
ContactUsLinkComponent
2,5-Dimethyl-3-nitropyridine 98%
2,5-Dimethyl-3-nitropyridine 98%
Catalog # APOSOR959822-1G
Supplier:  Apollo Scientific
CAS Number:  
2,5-Dimethyl-3-nitropyridine 98%
Catalog # APOSOR959822-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :152.151 g/mol
Storage Temperature:Ambient
CAS Number:52381-06-3
Formula:C₇H₈N₂O₂
MDL Number:MFCD10574716