Order Entry
Export
ContactUsLinkComponent
2,3-Cyclopentenopyridine N-oxide 98%
2,3-Cyclopentenopyridine N-oxide 98%
Catalog # APOSOR959735-1G
Supplier:  Apollo Scientific
CAS Number:  
2,3-Cyclopentenopyridine N-oxide 98%
Catalog # APOSOR959735-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :135.163 g/mol
Storage Temperature:Ambient
CAS Number:90685-58-8
Formula:C₈H₉NO
MDL Number:MFCD11110318