Order Entry
Export
ContactUsLinkComponent
5-Bromo-4-ethyl-3,4-dihydropyrimidine 97%
5-Bromo-4-ethyl-3,4-dihydropyrimidine 97%
Catalog # APOSOR959712-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Bromo-4-ethyl-3,4-dihydropyrimidine 97%
Catalog # APOSOR959712-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :189.053 g/mol
Storage Temperature:Ambient
CAS Number:224429-98-5
Formula:C₆H₉BrN₂
MDL Number:MFCD20681885