Order Entry
Export
ContactUsLinkComponent
2,5-Dimethoxypyridin-3-amine 95%
2,5-Dimethoxypyridin-3-amine 95%
Catalog # APOSOR959686-1G
Supplier:  Apollo Scientific
CAS Number:  
2,5-Dimethoxypyridin-3-amine 95%
Catalog # APOSOR959686-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :154.166 g/mol
Storage Temperature:Ambient
CAS Number:1211595-19-5
Formula:C₇H₁₀N₂O₂
MDL Number:MFCD09038237