Order Entry
Export
ContactUsLinkComponent
2-Oxa-6-azaspiro[3.3]heptane-6-ethanamine
2-Oxa-6-azaspiro[3.3]heptane-6-ethanamine
Catalog # APOSOR959446-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Oxa-6-azaspiro[3.3]heptane-6-ethanamine
Catalog # APOSOR959446-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :142.199 g/mol
Storage Temperature:Ambient
CAS Number:54384-40-6
Formula:C₇H₁₄N₂O
MDL Number:MFCD13184501