Order Entry
Export
Orders LinkContactUsLinkComponent
3,6-Dicyanopicoline 97%

3,6-Dicyanopicoline 97%

Supplier:   Apollo Scientific
3,6-Dicyanopicoline 97%
Supplier:  Apollo Scientific
Certificates
3,6-Dicyanopicoline 97%

2 Options Available Below

Product Details & Documents
  • CAS Number:1352318-05-8
  • Formula:C₈H₅N₃
  • MDL Number:MFCD20529525
  • Molecular Weight:143.145 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 189,00 €
    189,00 €
    1 * 250 mg
       189,00 €

  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 362,00 €
    362,00 €
    1 * 1 g
       362,00 €