Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Butyl-1,3-diazaspiro[4.4]non-1-en-4-one
2-Butyl-1,3-diazaspiro[4.4]non-1-en-4-one 95%
2-Butyl-1,3-diazaspiro[4.4]non-1-en-4-one 95%
:
Apollo Scientific
2-Butyl-1,3-diazaspiro[4.4]non-1-en-4-one 95%
:
Apollo Scientific
:
138402-05-8
:
:
C₁₁H₁₈N₂O
:
MFCD03411590
:
194.273 g/mol
:
Ambient