0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
2-Chloro-6-methylthiazolo[4,5-b]pyridine
2-Chloro-6-methylthiazolo[4,5-b]pyridine 95%
2-Chloro-6-methylthiazolo[4,5-b]pyridine 95%
:
Apollo Scientific
2-Chloro-6-methylthiazolo[4,5-b]pyridine 95%
:
Apollo Scientific
:
960535-45-9
:
:
C₇H₅ClN₂S
:
MFCD20528302
:
184.646 g/mol
:
GHS07
:
Ambient