0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
1-Methyl-2,3-dihydro-[1,1'-biphenyl]-4(1H)-one
1-Methyl-2,3-dihydro-[1,1'-biphenyl]-4(1H)-one 95%
1-Methyl-2,3-dihydro-[1,1'-biphenyl]-4(1H)-one 95%
1-Methyl-2,3-dihydro-[1,1'-biphenyl]-4(1H)-one 95%
Supplier:
Apollo Scientific
Certificates
Expand image
1-Methyl-2,3-dihydro-[1,1'-biphenyl]-4(1H)-one 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
17429-36-6
:
Formula:
C₁₃H₁₄O
MDL Number:
MFCD00087385
:
186.25 g/mol
Storage Temperature:
Ambient