Order Entry
Export
ContactUsLinkComponent
2,3-Dichloro-6-hydroxypyridine
Irritant
2,3-Dichloro-6-hydroxypyridine
Catalog # APOSOR958310-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,3-Dichloro-6-hydroxypyridine
Catalog # APOSOR958310-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :163.989 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:24525-63-1
Formula:C₅H₃Cl₂NO
MDL Number:MFCD16610775