Order Entry
Export
ContactUsLinkComponent
8-Bromoimidazo[1,2-a]pyrazine 97%
Irritant
8-Bromoimidazo[1,2-a]pyrazine 97%
Catalog # APOSOR957970-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
8-Bromoimidazo[1,2-a]pyrazine 97%
Catalog # APOSOR957970-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :198.02 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:69214-34-2
Formula:C₆H₄BrN₃
MDL Number:MFCD18072715