Order Entry
Export
ContactUsLinkComponent
2-Chloro-4-hydroxy-n,n-dimethylbenzamide 95%
2-Chloro-4-hydroxy-n,n-dimethylbenzamide 95%
Catalog # APOSOR957753-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Chloro-4-hydroxy-n,n-dimethylbenzamide 95%
Catalog # APOSOR957753-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :199.634 g/mol
Storage Temperature:Ambient
Formula:C₉H₁₀ClNO₂
MDL Number:MFCD16997928