Order Entry
Export
ContactUsLinkComponent
1-Hydroxy-2-methoxynaphthalene 95%
1-Hydroxy-2-methoxynaphthalene 95%
Catalog # APOSOR957697-1G
Supplier:  Apollo Scientific
CAS Number:  
1-Hydroxy-2-methoxynaphthalene 95%
Catalog # APOSOR957697-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :174.196 g/mol
Storage Temperature:Ambient
CAS Number:1888-41-1
Formula:C₁₁H₁₀O₂
MDL Number:MFCD14534802