Order Entry
Export
ContactUsLinkComponent
2,6-Dichloro-3-hydroxybenzonitrile 95%
2,6-Dichloro-3-hydroxybenzonitrile 95%
Catalog # APOSOR957691-1G
Supplier:  Apollo Scientific
CAS Number:  
2,6-Dichloro-3-hydroxybenzonitrile 95%
Catalog # APOSOR957691-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :188.011 g/mol
Storage Temperature:Ambient
CAS Number:3336-34-3
Formula:C₇H₃Cl₂NO
MDL Number:MFCD01662155