Order Entry
Export
ContactUsLinkComponent
 
4-(Trimethylsilyl)phenol 95%
Catalog # APOSOR957666-1G
Supplier:  Apollo Scientific
CAS Number:  
4-(Trimethylsilyl)phenol 95%
Catalog # APOSOR957666-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :166.292 g/mol
Storage Temperature:Ambient
CAS Number:13132-25-7
Formula:C₉H₁₄OSi
MDL Number:MFCD00034672