Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-(Trimethylsilyl)phenol
3-(Trimethylsilyl)phenol 95%
3-(Trimethylsilyl)phenol 95%
3-(Trimethylsilyl)phenol 95%
APOSOR957665-1G
:
Apollo Scientific
:
3-(Trimethylsilyl)phenol 95%
APOSOR957665-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
166.292 g/mol
:
Ambient
:
C₉H₁₄OSi
:
MFCD24713613