Order Entry
Export
ContactUsLinkComponent
3-Chloro-4,5-dimethoxyphenol 95%
3-Chloro-4,5-dimethoxyphenol 95%
Catalog # APOSOR957650-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Chloro-4,5-dimethoxyphenol 95%
Catalog # APOSOR957650-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :188.608 g/mol
Storage Temperature:Ambient
CAS Number:89981-66-8
Formula:C₈H₉ClO₃
MDL Number:MFCD24717705