Order Entry
Export
ContactUsLinkComponent
3-(2,2-Dimethylpropoxy)phenol 95%
3-(2,2-Dimethylpropoxy)phenol 95%
Catalog # APOSOR957636-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(2,2-Dimethylpropoxy)phenol 95%
Catalog # APOSOR957636-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :180.244 g/mol
Storage Temperature:Ambient
CAS Number:1394975-12-2
Formula:C₁₁H₁₆O₂
MDL Number:MFCD23111162