Order Entry
Export
ContactUsLinkComponent
3-Hydroxy-N-propylbenzamide 95%
3-Hydroxy-N-propylbenzamide 95%
Catalog # APOSOR957604-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Hydroxy-N-propylbenzamide 95%
Catalog # APOSOR957604-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :179.216 g/mol
Storage Temperature:Ambient
Formula:C₁₀H₁₃NO₂
MDL Number:MFCD11130580