Order Entry
Export
ContactUsLinkComponent
4-Ethoxy-2,5-dimethylphenol 95%
4-Ethoxy-2,5-dimethylphenol 95%
Catalog # APOSOR957597-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Ethoxy-2,5-dimethylphenol 95%
Catalog # APOSOR957597-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :166.217 g/mol
Storage Temperature:Ambient
CAS Number:99172-75-5
Formula:C₁₀H₁₄O₂
MDL Number:MFCD14537749