Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Hydroxy-3,6-dimethyl-2(1H)-pyridinone
4-Hydroxy-3,6-dimethyl-2(1H)-pyridinone 95%
4-Hydroxy-3,6-dimethyl-2(1H)-pyridinone 95%
4-Hydroxy-3,6-dimethyl-2(1H)-pyridinone 95%
Supplier:
Apollo Scientific
Certificates
Expand image
4-Hydroxy-3,6-dimethyl-2(1H)-pyridinone 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
4664-13-5
:
Formula:
C₇H₉NO₂
MDL Number:
MFCD11113422
:
139.152 g/mol
Storage Temperature:
Ambient