Order Entry
Export
ContactUsLinkComponent
4-Hydroxy-N-methoxybenzamide 95%
4-Hydroxy-N-methoxybenzamide 95%
Catalog # APOSOR957581-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Hydroxy-N-methoxybenzamide 95%
Catalog # APOSOR957581-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :167.162 g/mol
Storage Temperature:Ambient
Formula:C₈H₉NO₃
MDL Number:MFCD21101042