Order Entry
Export
ContactUsLinkComponent
3,5-Dichloro-4-methoxyphenol 97%
3,5-Dichloro-4-methoxyphenol 97%
Catalog # APOSOR957578-1G
Supplier:  Apollo Scientific
CAS Number:  
3,5-Dichloro-4-methoxyphenol 97%
Catalog # APOSOR957578-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :193.027 g/mol
Storage Temperature:Ambient
CAS Number:56680-68-3
Formula:C₇H₆Cl₂O₂
MDL Number:MFCD12405414