Order Entry
Export
ContactUsLinkComponent
4-Methoxy-3,5-dimethylphenol 95%
4-Methoxy-3,5-dimethylphenol 95%
Catalog # APOSOR957506-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Methoxy-3,5-dimethylphenol 95%
Catalog # APOSOR957506-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :152.19 g/mol
Storage Temperature:Ambient
CAS Number:4962-29-2
Formula:C₉H₁₂O₂
MDL Number:MFCD18432122