Order Entry
Export
ContactUsLinkComponent
5-Chloro-4-hydroxy-2-methoxypyridine 97%
5-Chloro-4-hydroxy-2-methoxypyridine 97%
Catalog # APOSOR957493-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Chloro-4-hydroxy-2-methoxypyridine 97%
Catalog # APOSOR957493-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :159.57 g/mol
Storage Temperature:Ambient
CAS Number:1196146-71-0
Formula:C₆H₆ClNO₂
MDL Number:MFCD13189878