Order Entry
Export
ContactUsLinkComponent
4-Chloro-3,5-dimethylanisole 95%
4-Chloro-3,5-dimethylanisole 95%
Catalog # APOSOR957064-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Chloro-3,5-dimethylanisole 95%
Catalog # APOSOR957064-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :170.636 g/mol
Storage Temperature:Ambient
CAS Number:6981-15-3
Formula:C₉H₁₁ClO
MDL Number:MFCD07780689