Order Entry
Export
ContactUsLinkComponent
4-Chloro-2,3-dimethoxypyridine 95%
4-Chloro-2,3-dimethoxypyridine 95%
Catalog # APOSOR957022-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Chloro-2,3-dimethoxypyridine 95%
Catalog # APOSOR957022-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :173.597 g/mol
Storage Temperature:Ambient
CAS Number:1261629-46-2
Formula:C₇H₈ClNO₂
MDL Number:MFCD18415676