Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Chloro-2,3-dihydrobenzo[b][1,4]dioxine
6-Chloro-2,3-dihydrobenzo[b][1,4]dioxine 95%
6-Chloro-2,3-dihydrobenzo[b][1,4]dioxine 95%
6-Chloro-2,3-dihydrobenzo[b][1,4]dioxine 95%
Supplier:
Apollo Scientific
Certificates
Expand image
6-Chloro-2,3-dihydrobenzo[b][1,4]dioxine 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
57744-68-0
:
Formula:
C₈H₇ClO₂
MDL Number:
MFCD16622016
:
170.593 g/mol
Storage Temperature:
Ambient