Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Spiro[3.5]nonane-1,3-dione
Spiro[3.5]nonane-1,3-dione ≥98%
Spiro[3.5]nonane-1,3-dione ≥98%
Spiro[3.5]nonane-1,3-dione ≥98%
Supplier:
Apollo Scientific
Certificates
Expand image
Spiro[3.5]nonane-1,3-dione ≥98%
Supplier:
Apollo Scientific
About this item
CAS Number:
455264-97-8
:
Formula:
C₉H₁₂O₂
MDL Number:
MFCD11110994
:
152.19 g/mol
Storage Temperature:
Ambient