Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Methyl-1,3-benzoxazol-2-amine
5-Methyl-1,3-benzoxazol-2-amine 98%
5-Methyl-1,3-benzoxazol-2-amine 98%
5-Methyl-1,3-benzoxazol-2-amine 98%
Supplier:
Apollo Scientific
Certificates
Expand image
5-Methyl-1,3-benzoxazol-2-amine 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
64037-15-6
:
Formula:
C₈H₈N₂O
MDL Number:
MFCD01664145
:
148.162 g/mol
Storage Temperature:
Ambient