Order Entry
Export
ContactUsLinkComponent
Methyl 2,5-dimethylthiophene-3-carboxylate 95%
Methyl 2,5-dimethylthiophene-3-carboxylate 95%
Catalog # APOSOR955944-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 2,5-dimethylthiophene-3-carboxylate 95%
Catalog # APOSOR955944-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :170.229 g/mol
Storage Temperature:Ambient
CAS Number:53562-48-4
Formula:C₈H₁₀O₂S
MDL Number:MFCD23714100