Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,6-Dichloro-2-methylaniline
3,6-Dichloro-2-methylaniline 95%
3,6-Dichloro-2-methylaniline 95%
3,6-Dichloro-2-methylaniline 95%
Supplier:
Apollo Scientific
Certificates
Expand image
3,6-Dichloro-2-methylaniline 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
62077-26-3
:
Formula:
C₇H₇Cl₂N
MDL Number:
MFCD20699076
:
176.043 g/mol
Storage Temperature:
Ambient