Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,4,7-Trimethyl-1,3-dihydroquinolin-2-one
4,4,7-Trimethyl-1,3-dihydroquinolin-2-one 97%
4,4,7-Trimethyl-1,3-dihydroquinolin-2-one 97%
4,4,7-Trimethyl-1,3-dihydroquinolin-2-one 97%
Supplier:
Apollo Scientific
Certificates
Expand image
4,4,7-Trimethyl-1,3-dihydroquinolin-2-one 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
133999-05-0
:
Formula:
C₁₂H₁₅NO
MDL Number:
MFCD26131173
:
189.254 g/mol
Storage Temperature:
Ambient