Order Entry
Export
ContactUsLinkComponent
2,3-Dimethylindoline 95%
2,3-Dimethylindoline 95%
Catalog # APOSOR954291-1G
Supplier:  Apollo Scientific
CAS Number:  
2,3-Dimethylindoline 95%
Catalog # APOSOR954291-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :147.217 g/mol
Storage Temperature:Ambient
CAS Number:22120-50-9
Formula:C₁₀H₁₃N
MDL Number:MFCD00005707