Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-Methyl-3,4-dimethylbenzylamine
N-Methyl-3,4-dimethylbenzylamine 95%
N-Methyl-3,4-dimethylbenzylamine 95%
N-Methyl-3,4-dimethylbenzylamine 95%
Supplier:
Apollo Scientific
Certificates
Expand image
N-Methyl-3,4-dimethylbenzylamine 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
165741-71-9
:
Formula:
C₁₀H₁₅N
MDL Number:
MFCD11204739
:
149.233 g/mol
Storage Temperature:
Ambient