Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Oxaspiro[2.5]octane
1-Oxaspiro[2.5]octane 97%
1-Oxaspiro[2.5]octane 97%
1-Oxaspiro[2.5]octane 97%
Supplier:
Apollo Scientific
Certificates
Expand image
1-Oxaspiro[2.5]octane 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
185-70-6
:
Formula:
C₇H₁₂O
MDL Number:
MFCD00022348
:
112.17 g/mol
Storage Temperature:
Ambient